Learn More
Molex Molex-Polymicro Capillary 40UMx105UM 10M Reel
Supplier:  Molex 1068150596/10M
Encompass_Preferred
TSP040105 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 20um ID x 90um OD, spool length of 10m.
Catalog No. 50-110-7676