Learn More
Molex Molex-Polymicro Capillary 40UMx105UM 25M Reel
Catalog No. 501108596
Encompass_Preferred
TSP040105 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 105um OD, spool length of 25m.
Catalog No. 50-110-8596 Supplier Molex Supplier No. 1068150596/25M
TSP040105 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 105um OD, spool length of 25m.