Learn More
Molex Molex-Polymicro Capillary 40UMx150UM 25M Reel
Catalog No. 501108597
Encompass_Preferred
TSP040150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 150um OD, spool length of 25m.
Catalog No. 50-110-8597 Supplier Molex Supplier No. 1068150014/25M
TSP040150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 150um OD, spool length of 25m.