Learn More
Molex Molex-Polymicro Capillary 40UMx363UM 25M Reel
Catalog No. 501108598
Encompass_Preferred
TSP040375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8598 Supplier Molex Supplier No. 1068150383/25M
TSP040375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 363um OD, spool length of 25m.