Learn More
Molex Molex-Polymicro Capillary 50UMx363UM 25M Reel
Catalog No. 501108600
Encompass_Preferred
TSP050375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 50um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8600 Supplier Molex Supplier No. 1068150017/25M
TSP050375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 50um ID x 363um OD, spool length of 25m.