Learn More
Molex Molex-Polymicro Capillary 50UMx363UM 50M Reel
Catalog No. 501108620
Encompass_Preferred
TSP050375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 50um ID x 363um OD, spool length of 50m.
Catalog No. 50-110-8620 Supplier Molex Supplier No. 1068150017/50M
TSP050375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 50um ID x 363um OD, spool length of 50m.