Learn More
Thermo Scientific CAPILLARY 65M X.5ID
Supplier:  Thermo Scientific 7011120100
Encompass
LC/MS Instrument Parts; Ion transfer tube for Quantum, Quantum AM and Quantum Discovery; Thermo Scientific; Replacement part for TSQ Quantum Triple Quadrupole LC/MS
Catalog No. NC9879465