Learn More
Molex Molex-Polymicro Capillary 75UMx150UM 10M Reel
Supplier:  Molex 1068150018/10M
Encompass_Preferred
TSP075150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 150um OD, spool length of 10m.
Catalog No. 50-110-7681