Learn More
Molex Molex-Polymicro Capillary 75UMx150UM 25M Reel
Catalog No. 501108601
Encompass_Preferred
TSP075150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 150um OD, spool length of 25m.
Catalog No. 50-110-8601 Supplier Molex Supplier No. 1068150018/25M
TSP075150 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 150um OD, spool length of 25m.