Learn More
Molex Molex-Polymicro Capillary 75UMx363UM 25M Reel
Catalog No. 501108602
Encompass_Preferred
TSP075375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 363um OD, spool length of 25m.
Catalog No. 50-110-8602 Supplier Molex Supplier No. 1068150019/25M
TSP075375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 75um ID x 363um OD, spool length of 25m.