Learn More
MilliporeSigma™ Supelco™ Capillary Cleaving™ Tools and Accessories
Shop All MilliporeSigma Supelco ProductsDescription
- Makes scalpel-like cuts to polyimide-coated fused silica
- Leaves no jagged edges to create problems
- Industrial sapphire cutting edges remains sharp indefinitely
- Spring-loaded retractable blade reduces the chance of breakage if the tool is dropped
- Replacement sapphire cutting blades are available
Specifications
Specifications
| For Use With (Application) | Making Scalpel-Like Cuts to Polyimide-Coated Fused Silica |
| Material | Sapphire Cutting Blade |
| Product Type | Fixed Blade Version |
Cleaving is a trademark of Sigma-Aldrich Co. LLC