Learn More
Agilent Technologies CAPILLARY CONDITIONING SOLUTIO
Catalog No. NC9949677
Encompass
Capillary Conditioning Solution, 1000 mL. Capillary conditioning solution for use with Oligel or Native Oligel Plus Gel Matrix on the Oligo PRO parallel capillary electrophoresis system (Advanced Anaytical Technologies, Inc.).
Catalog No. NC9949677 Supplier Agilent Technologies Supplier No. DN4751000
Capillary Conditioning Solution, 1000 mL. Capillary conditioning solution for use with Oligel or Native Oligel Plus Gel Matrix on the Oligo PRO parallel capillary electrophoresis system (Advanced Anaytical Technologies, Inc.).