Learn More
Instech Capillary / dispensing tube, 50µL, K3EDTA, violet (Sarstedt Minivette POCT 17.2113.150)
SDP
Catalog No. NC2123271
Encompass
Sarstedt’s capillary tubes streamline the collection of small blood samples. When combined with Instech Vascular Access Buttons™ or PinPorts™ they can be part of a significant refinement in rodent blood collection.
Capillary / dispensing tubes (Minivette POCT) collect precise 10, 20, 50, 100 or 200µL samples, with or without an anticoagulant, which are then easily transferred for analysis without a syringe.
The PNP3MS connector easily presses onto the tubes and then punctures the small septum on Vascular Access Buttons™ and PinPorts™ to draw up the sample. You can take samples directly from a catheterized animal, or for completely hands off sampling, at the top of the cage. Arterial pressure is required for best results (for example, from a carotid artery catheterization).
For laboratory animal research use only.
This item is not returnable. View return policy
Catalog No. NC2123271 Supplier Instech Supplier No. CTPOCT050K3EDTA
Sarstedt’s capillary tubes streamline the collection of small blood samples. When combined with Instech Vascular Access Buttons™ or PinPorts™ they can be part of a significant refinement in rodent blood collection.
Capillary / dispensing tubes (Minivette POCT) collect precise 10, 20, 50, 100 or 200µL samples, with or without an anticoagulant, which are then easily transferred for analysis without a syringe.
The PNP3MS connector easily presses onto the tubes and then punctures the small septum on Vascular Access Buttons™ and PinPorts™ to draw up the sample. You can take samples directly from a catheterized animal, or for completely hands off sampling, at the top of the cage. Arterial pressure is required for best results (for example, from a carotid artery catheterization).
For laboratory animal research use only.