Learn More
Microsolv Technology Corporation Capillary Fused Silica 50Um
Catalog No. 5001452696
Encompass_Preferred
Capillary, Bare Fused Silica, 50um ID by 1 meter. Simplus Brand. 1/EA.
Catalog No. 50-014-52696 Supplier Microsolv Technology Corporation Supplier No. 04050C
Capillary, Bare Fused Silica, 50um ID by 1 meter. Simplus Brand. 1/EA.