Learn More
Microsolv Technology Corporation Capillary Fused Silica 75Um 25
Catalog No. 5001452701
Encompass_Preferred
Capillary, Bare Fused Silica, 75um ID by 25 meters. Simplus Brand. 1/EA.
Catalog No. 50-014-52701 Supplier Microsolv Technology Corporation Supplier No. 04071C25
Capillary, Bare Fused Silica, 75um ID by 25 meters. Simplus Brand. 1/EA.