Learn More
Perkin Elmer US LLC Capillary Injector with PPC, 240 V
Supplier:  Perkin Elmer US LLC N6120139
Encompass_Preferred
The AutoSystem XL must be PPC (Programmable Pneumatic Control) ready. If not, a PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.
Catalog No. 50-211-3736
Add to Cart
Request a Quote