Learn More
Analytical Sales & Services Capillary PEEK Tubing - 1/32" OD x 175µm IDx 5' Length
Catalog No. 5000804837
Encompass
Capillary PEEK Tubing \n\n\n\nOuter Diameter: 1/32"\n\nInner Diameter: 175µm (0.007")\n\nLength: 5'\n\nColor: Yellow\n\nPressure Rating: 5,000psi (345 bar)
Catalog No. 50-008-04837 Supplier Analytical Sales & Services Supplier No. 10575
Capillary PEEK Tubing \n\n\n\nOuter Diameter: 1/32"\n\nInner Diameter: 175µm (0.007")\n\nLength: 5'\n\nColor: Yellow\n\nPressure Rating: 5,000psi (345 bar)