Learn More
Analytical Sales & Services Capillary PEEK Tubing - 1/32" OD x 250µm ID x 5' Length
Catalog No. 5000804973
Encompass
Capillary PEEK Tubing\n\n\n\nOuter Diameter: 1/32"\n\nInner Diameter: 250µm (0.010")\n\nLength: 5'\n\nColor: Blue\n\nPressure Rating: 5,000psi (345 bar)
Catalog No. 50-008-04973 Supplier Analytical Sales & Services Supplier No. 10585
Capillary PEEK Tubing\n\n\n\nOuter Diameter: 1/32"\n\nInner Diameter: 250µm (0.010")\n\nLength: 5'\n\nColor: Blue\n\nPressure Rating: 5,000psi (345 bar)