Learn More
Analytical Sales & Services Capillary PEEK Tubing - 360µm OD x 150µm ID x 5' Length
Catalog No. 5000806690
Encompass
Capillary PEEK Tubing - 360µm OD x 150µm ID x 5' Length Color: Yellow Pressure Rating: 2,000psi (138 bar)
Catalog No. 50-008-06690 Supplier Analytical Sales & Services Supplier No. 1080
Capillary PEEK Tubing - 360µm OD x 150µm ID x 5' Length Color: Yellow Pressure Rating: 2,000psi (138 bar)