Learn More
Analytical Sales & Services Capillary PEEK Tubing - 360µm OD x 50µm ID x 5' Length
Catalog No. 5000806977
Encompass
Capillary PEEK Tubing - 360µm OD x 50µm ID x 5' Length Color: Natural Pressure Rating: 2,000psi (138 bar)
Catalog No. 50-008-06977 Supplier Analytical Sales & Services Supplier No. 1050
Capillary PEEK Tubing - 360µm OD x 50µm ID x 5' Length Color: Natural Pressure Rating: 2,000psi (138 bar)