Learn More
Nanotemper Technologies Inc NanoTemper Technologies SKU: PR-P006, Capillary Remover
Catalog No. NC3780025
Encompass
Capillary Remover for use with the Prometheus NT.48. The Capillary Remover is designed for removing capillaries from the Prometheus NT.48 instrument after the measurement in one scoop to minimize handling steps.
Catalog No. NC3780025 Supplier Nanotemper Technologies Inc Supplier No. PRP006
Capillary Remover for use with the Prometheus NT.48. The Capillary Remover is designed for removing capillaries from the Prometheus NT.48 instrument after the measurement in one scoop to minimize handling steps.