Learn More
Perkin Elmer US LLC Capillary Sample Tubes, 100/pk
Catalog No. 502113339
Encompass_Preferred
For use in direct sample analysis (DSA). Borosilicate glass capillary tubes, 14.5 mm in length.
Catalog No. 50-211-3339 Supplier Perkin Elmer US LLC Supplier No. MZ305385
For use in direct sample analysis (DSA). Borosilicate glass capillary tubes, 14.5 mm in length.