Learn More
Electron Microscopy Sciences Borosilicate Capillary Glass Slide, 0.20x2.0 mm, 100 mm, 0.150 Wall thick
SDP
Catalog No. 5033548
Encompass_Preferred
Micro glass slides are precision rectangular glass capillary tubes for use in microscopic viewing of liquid samples. Capillary action instantly collects the sample and the thin glass walls minimize evaporation. The design of these cells eliminates the needs for cover glass and the hanging drop method.
Made from Corning 7740 borosilicate, low expansion, type I glass, these micro glass slides can withstand heat up to 500 degree C. Samples can be retained for future reference by sealing or cementing open ends.
Micro glass slide are packaged in plastic vials.
I.D. tolerance +/- 10% and wall tolerance +/- 20%.
I.D.: 0.20 x 2.0 mm
Length: 100 mm
Wall thickness: 0.150
Catalog No. 50-335-48 Supplier Electron Microscopy Sciences Supplier No. 6382510
Micro glass slides are precision rectangular glass capillary tubes for use in microscopic viewing of liquid samples. Capillary action instantly collects the sample and the thin glass walls minimize evaporation. The design of these cells eliminates the needs for cover glass and the hanging drop method.
Made from Corning 7740 borosilicate, low expansion, type I glass, these micro glass slides can withstand heat up to 500 degree C. Samples can be retained for future reference by sealing or cementing open ends.
Micro glass slide are packaged in plastic vials.
I.D. tolerance +/- 10% and wall tolerance +/- 20%.
I.D.: 0.20 x 2.0 mm
Length: 100 mm
Wall thickness: 0.150