Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Capillary Transfer Pipette
GREENER_CHOICE

Catalog No. 02707031
Change view
Click to view available options
Volume (Metric) Dispense:
25 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Dispense
02-707-031 50 μL
02-707-030 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-707-031 Supplier Thermo Scientific™ Supplier No. 1029100
Only null left
Add to Cart
Add to Cart

With an innovative design, Thermo Scientific™ Samco capillary transfer pipettes are well suited for collection and dispensing of microliter-scale liquid samples.

With its innovative design, Thermo Scientific™ Samco™ Capillary Transfer Pipettes are suited for the collection and dispensing of micro samples of liquid. This makes them ideal for a wide range of rapid tests. The pipettes use capillary action to automatically draw the liquid for precise and accurate sample collection.

  • Ideal for kitting
  • Uniquely designed with durable, yet soft plastic, helping to reduce the worry of breakage
  • Accurately dispense 50 μL, with customization options available
  • Single-use
  • Dispenses liquid within seconds without a plunger
  • No dripping during the transfer stage
  • No anticoagulant coating required; collects and dispenses liquid within seconds
  • Easy-to-see volume marking, allowing for accurate dispensing
  • Fills effortlessly without air bubbles
  • Made from safe, nontoxic materials – free of latex, DEHP, and BPA

Compliance

Meets ISO 9001-2008 and 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 2.34 mm
Material Polyethylene
Sterility Non-sterile
Quantity 5000/Cs.
Volume (Metric) Dispense 50 μL
Packaging Type 100 per bag, 5 bags per box, 10 boxes per case
Color Clear
Volume (Metric) 50 μL
Length (Metric) 3.87 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.