Learn More
Optimize Technologies OPTI-TRAP™ Capillary Trap Cartridge, SCX, 0.5µL, 0.5mm
SDP
Catalog No. 502006162
Encompass_Preferred
The OPTI-TRAP™is a bi-directional trap cartridge system which is used individually or in a series for sample concentration and purification. The OPTI-TRAP™ system features a durable biocompatible trap holder assembly made of either PEEK (capillary configuration) or stainless steel with a PEEK flow path (micro and macro configurations). OPTI-TRAPs feature low back pressure and are loaded and eluted manually with a syringe or on-line when plumbed into the sample injection valve. Additionally OPTI-TRAPs include titanium frits for full biocompatibility and custom packing is also available upon request. The cartridges are available in three different sizes: 50uL, 5.0uL & 0.5uL and can be purchased as singles or in 6 packs.
Catalog No. 50-200-6162 Supplier Optimize Technologies Supplier No. 1004813TP
The OPTI-TRAP™is a bi-directional trap cartridge system which is used individually or in a series for sample concentration and purification. The OPTI-TRAP™ system features a durable biocompatible trap holder assembly made of either PEEK (capillary configuration) or stainless steel with a PEEK flow path (micro and macro configurations). OPTI-TRAPs feature low back pressure and are loaded and eluted manually with a syringe or on-line when plumbed into the sample injection valve. Additionally OPTI-TRAPs include titanium frits for full biocompatibility and custom packing is also available upon request. The cartridges are available in three different sizes: 50uL, 5.0uL & 0.5uL and can be purchased as singles or in 6 packs.