Learn More
Waters Corp Waters Corporation Capillary Tube, Stainless Steel, ESI Probes, 2 per Pack
Catalog No. 50792955 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement steel capillary is designed for original ESI probes used on early Xevo sourced Mass Spectrometers. It is supplied as a 2 pk.
  • Material: Stainless steel
  • Units per package: 2 pk
  • Type: Tubing
  • System type: MS
  • Designed for original ESI probes
  • Replacement schedule: As required
Catalog No. 50-792-955 Supplier Waters Corp Supplier No. 700004206
This replacement steel capillary is designed for original ESI probes used on early Xevo sourced Mass Spectrometers. It is supplied as a 2 pk.
  • Material: Stainless steel
  • Units per package: 2 pk
  • Type: Tubing
  • System type: MS
  • Designed for original ESI probes
  • Replacement schedule: As required