Learn More
Thermo Scientific CAPILLARY TUBE
Supplier:  Thermo Scientific 9705520198
Encompass
Ion transfer tube replaceable for LTQFT, LTQFT Ultra and Orbitrap spectrometers 550a�m Thermo Scientific Hypersil by Thermo Scientific Chromatography Consumable.
Catalog No. NC9135296