Learn More
Instech PinPort-to-Sarstedt capillary tube connector, 50 sterile pouches of 5 (250 pcs)
SDP
Catalog No. NC2122960
Encompass
This PNP3MS connector easily presses onto both capillary / centrifugation tubes (Microvette CB 300) and capillary / dispensing tubes (Minivette POCT) . The connector then punctures the small septum on Vascular Access Buttons™ and PinPorts™ to draw up the sample. You can take samples directly from a catheterized animal, or for completely hands off sampling, at the top of the cage. Arterial pressure is required for best results (for example, from a carotid artery catheterization).
Sarstedt’s capillary tubes streamline the collection of small blood samples. When combined with Instech Vascular Access Buttons™ or PinPorts™ they can be part of a significant refinement in rodent blood collection.
For laboratory animal research use only.
This item is not returnable. View return policy
Catalog No. NC2122960 Supplier Instech Supplier No. PNP3MS
This PNP3MS connector easily presses onto both capillary / centrifugation tubes (Microvette CB 300) and capillary / dispensing tubes (Minivette POCT) . The connector then punctures the small septum on Vascular Access Buttons™ and PinPorts™ to draw up the sample. You can take samples directly from a catheterized animal, or for completely hands off sampling, at the top of the cage. Arterial pressure is required for best results (for example, from a carotid artery catheterization).
Sarstedt’s capillary tubes streamline the collection of small blood samples. When combined with Instech Vascular Access Buttons™ or PinPorts™ they can be part of a significant refinement in rodent blood collection.
For laboratory animal research use only.