Learn More
Thomas Scientific Capillary Tube, Micro-hematocrit, Soda Lime Glass, Red Tip, Sodium Heparinized, 1000/cs
SDP
Catalog No. NC1740821
Encompass
These glass capillary tubes are designed for both safe blood collection as well as accurate micro-hematocrit determinations. Precision molded, they provide excellent sample retention during centrifugation. In addition, the tubes fill rapidly without affecting specimen integrity.
Catalog No. NC1740821 Supplier Thomas Scientific Supplier No. 1202K16
These glass capillary tubes are designed for both safe blood collection as well as accurate micro-hematocrit determinations. Precision molded, they provide excellent sample retention during centrifugation. In addition, the tubes fill rapidly without affecting specimen integrity.