Learn More
Capillary Tubes
SCI_ED
Catalog No. S37609E
Change view
Click to view available options
90mm Melting-point capillary
Catalog No.
S37609E 90mm Melting-point capillary
1 options
This item is not returnable. View return policy
Catalog No. S37609E Supplier Friedrich & Dimmock Inc Supplier No. 121415/EMD
This item is not returnable. View return policy
Indispensible for calculating blood clotting time or determining melting points.
  • Made of alkali-free, corrosion-resistant glass
  • Extra-thin walls break easily and cleanly
  • Good for routine melting point determinations Capacity: 30μL One sealed end
  • 90mm melting-point capillary
  • Sold 100 tubes per vial
Specifications
Description One end sealed
Material Glass
Type Capillary Tube