Learn More
Phenomenex Inc Capillary Tubing, PEEK
Catalog No. 502400630 Shop All Phenomenex Products
Encompass_Preferred
Capillary Tubing, PEEK 5ft L x 1/16in OD x 0.010in ID, Blue
Catalog No. 50-240-0630 Supplier Phenomenex Inc Supplier No. AT01107
Capillary Tubing, PEEK 5ft L x 1/16in OD x 0.010in ID, Blue