Learn More
Waters Corp Waters Corporation Tubing Assembly, Capillary, With Frit, 40 μm, 16 in.
Catalog No. 50791931 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing with frit is designed for use with the 10,000psi nanoACQUITY system. It connects from the nano tee to the trap valve tube, supporting various system configurations for optimized performance.
  • Designed for 10,000psi nanoACQUITY systems.
  • Connects nano tee to the trap valve tube.
  • Supports direct injection configurations.
  • Supports 2D with dilution configurations.
  • Supports single pump trapping configurations.
  • Supports vented dual pump trapping configurations.
  • Includes integrated frit.
Catalog No. 50-791-931 Supplier Waters Corp Supplier No. 430001571
This capillary tubing with frit is designed for use with the 10,000psi nanoACQUITY system. It connects from the nano tee to the trap valve tube, supporting various system configurations for optimized performance.
  • Designed for 10,000psi nanoACQUITY systems.
  • Connects nano tee to the trap valve tube.
  • Supports direct injection configurations.
  • Supports 2D with dilution configurations.
  • Supports single pump trapping configurations.
  • Supports vented dual pump trapping configurations.
  • Includes integrated frit.