Learn More
Andwin Scientific CAPILLRY/PISTN 1000UL 180/3
SDP
Supplier:  Andwin Scientific C1000/DEL
Encompass
The Positive Displacement tips are presterilized and preassembled. The tips have a maximum volume capacity of 1000 L and have capillaries with pistons tip technology. This product is specially designed to be compatible with Pos-D and Gilson Microman positive-displacement pipettes.
Catalog No. NC0018120