Learn More
3M™ DBI-Sala™ Fixed I-Beam Anchor
Catalog No. 19066851 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
2.5 to 18 W x 1.5 in. Thick beam flange
2.5 to 24 W x 1.5 in. Thick beam flange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-066-851 2.5 to 24 W x 1.5 in. Thick beam flange
19-066-850 2.5 to 18 W x 1.5 in. Thick beam flange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-066-851 Supplier 3M™ Supplier No. 7100241958
Add to Cart
Add to Cart
Easily adjustable in any orientation or direction
  • Anchor is easily installed by sliding the adjustable hook-end sown to the flange and securing a pin
  • Installs in seconds to the beam structure
  • Built-in adjustment handle for final tightening
  • Attach it to the beam structure in any orientation or direction for added versatility
  • 2268 kg (5000 lb.) maximum strength in any direction
  • Temporarily fixed
  • With D ring
  • Provides economical flexibility
Compliance
Meets OSHA 1910.66 and OSHA 1926.502; ANSI™ A10.32 and ANSI Z359.1 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Includes D-ring
Strength 5000 lbf.
For Use With (Equipment) 2.5 to 24 W x 1.5 in. Thick beam flange
Certifications/Compliance OSHA, ANSI, ANSI Z359
Product Type Fixed beam anchor