Learn More
3M™ DBI-SALA™ Nano-Lok™ Self Retracting Lifeline
Catalog No. 19810555 Shop All 3M Products
Change view
Click to view available options
Description:
6 ft. (1.8 m) of 3/4 in. (19 mm) Dyneema™ fiber and polyester web and steel rebar hook, swiveling anchor loop.
6 ft. (1.8 m) of 3/4 in. (19 mm) Dynema polyester web and snap hook, swiveling anchor loop.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-810-555 6 ft. (1.8 m) of 3/4 in. (19 mm) Dyneema™ fiber and polyester web and steel rebar hook, swiveling anchor loop.
19-810-554 6 ft. (1.8 m) of 3/4 in. (19 mm) Dynema polyester web and snap hook, swiveling anchor loop.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-810-555 Supplier 3M™ Supplier No. 7000127176
Dyneema™ web and hook with loop.
  • Swiveling anchorage loop
  • Extremely compact, lightweight design
  • Tough and impact resistant thermoplastic housing
  • Quick activating speed sensing brake system
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 2.9 lb.
Description 6 ft. (1.8 m) of 3/4 in. (19 mm) Dyneema™ fiber and polyester web and steel rebar hook, swiveling anchor loop.
Length (English) 6 ft.
Certifications/Compliance ANSI A10.32, OSHA 1926.502, OSHA 1910.66, ANSI Z359.1, ANSI Z359.14
Product Type Self-retracting Lifeline - Web