Learn More
3M™ DBI-Sala™ Order Picker Fall Protection System
Catalog No. 19809361 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Ideal for mounted on top/bottom of cab with self retracting lifeline faced up
Catalog No. For Use With (Application)
19-809-361 Ideal for mounted on top/bottom of cab with self retracting lifeline faced up
1 options
Catalog No. 19-809-361 Supplier 3M™ Supplier No. 3103120
With reserve lifeline retention system for added safety
  • Cab-mount bracket with all fastening hardware
  • Stainless-steel components for corrosion resistance
  • Self locking swivel snap hook with impact indicator
  • Swiveling anchorage loop prevents twisting of lifeline
  • Can be mounted on top/bottom of cab with self retracting lifeline (SRL) faced up or down
  • 11.1 ft. (3.35m) nylon web
  • Weight: 15 lb. (6.8kg)
Compliance
Meets ANSI™ Z359.14-2012 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel, Nylon
Weight (Metric) 6.8kg
Weight (English) 15 lb.
Certifications/Compliance OSHA, ANSI, ANSI Z359
For Use With (Application) Ideal for mounted on top/bottom of cab with self retracting lifeline faced up