Learn More
3M™ DBI-Sala™ Sayflink Sleeves for Multi-Span Sayfline System
Catalog No. 191203585 Shop All 3M Products
Change view
Click to view available options
Certifications/Compliance:
OSHA , ANSI
Catalog No. Certifications/Compliance
19-120-3585 OSHA , ANSI
1 options
Catalog No. 19-120-3585 Supplier 3M™ Supplier No. 7100301368
Add to Cart
Add to Cart
Unique one-piece sleeve easily slides over the intermediate brackets with no manipulation required by the user for hands-free operation
  • When designing Sayfline horizontal lifeline system, Sayflink sleeves are ordered separately
  • Maximum of two Sayflink sleeves used per system
Compliance
Meets or exceeds all applicable industry standards including OSHA and ANSI.
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 0.29kg
Weight (English) 0.65 lb.
For Use With (Equipment) Multi span horizontal lifeline system
For Use With (Application) Ideal for multitude, hands-free operation
Certifications/Compliance OSHA , ANSI