Learn More
3M™ DBI-Sala™ Talon™ Self-Retracting Lifelines
Catalog No. 19066833 Shop All 3M Products
Change view
Click to view available options
Includes:
Cab mount bracket, self locking swivel snap hook
Snap locking hook
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-066-833 Snap locking hook
19-809-356 Cab mount bracket, self locking swivel snap hook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-066-833 Supplier 3M™ Supplier No. 3101000
Engineered for reliable protection with automatic lock and unlock system
  • Designed for smooth consistent operation
  • Lightweight design (3 lb.) and quick-connect system gives freedom of movement without compromising safety
  • Due to small size it can be connected directly to back of harness for added mobility
  • Compact form with order picker handle
  • All metal braking system for durability
  • Self-locking swivel snap hook with impact indicator
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and stringent ANSI Z359
TRUSTED_SUSTAINABILITY
Specifications
Includes Snap locking hook
Length (English) 8 ft.
Length (Metric) 2.43 m
Product Type Self-retracting lifeline
Certifications/Compliance OSHA, ANSI, ANSI Z359