Learn More
Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kit
GREENER_CHOICE
Catalog No. 14377801 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
14-377-801 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 14-377-801 Supplier Thermo Scientific™ Supplier No. 90111
  • $283.00
    $258.65 / Case of 100
    Save $24.35
    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Collect, store and transport urine samples for drugs of abuse testing with Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits.
Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits provide a collection beaker, transport vial, and specimen transport bag in a tamper evident package to support drug testing programs. The kits are constructed from a low-binding polypropropylene resin that has been laboratory tested for interaction with drugs of abuse analytes.
  • Easy-to-use—simply peel away the foil-top lid, remove the components and provide donor with the collection cup
  • After specimen is collected, transfer urine into the tamper-evident vial, close the hinged lid until it snaps, and bag for transport
Ordering Information:
Case of 100. Custom branding available for foil tops and specimen bags.
Includes:
Collection cup (8 oz.) with temperature indicator strip, 45mL Tamper evident transport vial, dual pocket specimen bag with absorbent pad, and a foil top seal
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection Kit
Material Polypropylene
Sterility Sterile
Capacity (Metric) 45 mL
Description Single Specimen Collection Kit
Includes Collection cup (8 oz.) with temperature indicator strip, 45 mL Tamper evident transport vial, Generic dual pocket specimen bag, Absorbent pad and Foil top
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.