Learn More
Medchemexpress LLC HY-15431 50mg , Capivasertib CAS:1143532-39-1 Purity:>98%
SDP
Catalog No. 502259255
Encompass_Preferred
HY-15431 50mg Capivasertib CAS: 1143532-39-1 Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9255 Supplier Medchemexpress LLC Supplier No. HY1543150MG
HY-15431 50mg Capivasertib CAS: 1143532-39-1 Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.