Learn More
Abcam Capivasertib (AZD5363), 100MG
Catalog No. 502671531
Encompass_Preferred
MW 428.9 Da, Purity >95%. A novel pyrrolopyrimidine derivative. A potent inhibitor of all AKT isoforms, IC₅₀ <10 nM, with potential antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1531 Supplier Abcam Supplier No. AB282438100MG
MW 428.9 Da, Purity >95%. A novel pyrrolopyrimidine derivative. A potent inhibitor of all AKT isoforms, IC₅₀ <10 nM, with potential antineoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement