Learn More
Selleck Chemical LLC Capivasertib (AZD5363) 5mg 1143532-39-1
SDP
Supplier:  Selleck Chemical LLC S80195MG
Encompass_Preferred
Capivasertib (AZD5363) potently inhibits all isoforms of Akt(Akt1/Akt2/Akt3) with IC50 of 3 nM/8 nM/8 nM in cell-free assays, similar to P70S6K/PKA and lower activity towards ROCK1/2. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7955