Learn More
Research Products International Corp Capless Micro-Tubes, Natural, 1.5ml, 500 per Case
SDP
Catalog No. 50998561
Encompass
Capless microcentrifuge tubes are useful for a variety of laboratory applications. Ideal for centrifuging, collecting fractions, mixing reagents and as holders for smaller size tubes. Tubes feature graduations and frosted writing area. Manufactured of a homopolymer that withstands high speed centrifugation and boiling applications.
Catalog No. 50-998-561 Supplier Research Products International Corp Supplier No. 145657
Capless microcentrifuge tubes are useful for a variety of laboratory applications. Ideal for centrifuging, collecting fractions, mixing reagents and as holders for smaller size tubes. Tubes feature graduations and frosted writing area. Manufactured of a homopolymer that withstands high speed centrifugation and boiling applications.