Learn More
Alkali Scientific CapLock Clip, for 5mL Centrifuge Tubes, with Breakaway Lifting Tabs, Maintains Closure Seal at High Temperatures, 20/cs
SDP
Catalog No. 502389129
Encompass_Preferred
CapLock Clip, with breakaway lifting tabs, maintains closure seal at high temperatures. Twenty per pack.
Catalog No. 50-238-9129 Supplier Alkali Scientific Supplier No. C2586
CapLock Clip, with breakaway lifting tabs, maintains closure seal at high temperatures. Twenty per pack.