Learn More
Andwin Scientific CAPLOCKS FOR 1.5ML TUBES
SDP
Catalog No. NC9638461
Encompass_Preferred
The cap locks by Andwin Scientific provide reliability and security by preventing the caps of 1.5 mL tubes to pop open. This product comes in a pack of 100.
Catalog No. NC9638461 Supplier Andwin Scientific Supplier No. 46658
The cap locks by Andwin Scientific provide reliability and security by preventing the caps of 1.5 mL tubes to pop open. This product comes in a pack of 100.