Learn More
Trajan Scientific CAPLRY CLMN 6QC5/HT5-0.10
SDP
Catalog No. 502530236
Encompass_Preferred
SGE HT5 Capillary Polyimide-Clad Column, Diameter Inner: 0.53 mm, Length: 6 m, Film Thickness: 0.1 um, For Use With: Pesticide Analysis, Herbicides Analysis, GC, Packing Material: SGE HT5, Particle Size: 0.1 um, Stationary Phase: 5% Phenyl (equiv.) Polycarborane Siloxane
Catalog No. 50-253-0236 Supplier Trajan Scientific Supplier No. 054655
SGE HT5 Capillary Polyimide-Clad Column, Diameter Inner: 0.53 mm, Length: 6 m, Film Thickness: 0.1 um, For Use With: Pesticide Analysis, Herbicides Analysis, GC, Packing Material: SGE HT5, Particle Size: 0.1 um, Stationary Phase: 5% Phenyl (equiv.) Polycarborane Siloxane