Learn More
MSC Caplugs Aluminum Foil Masking Discs, 1/4 in Diameter, Silver, 2000/Roll
Catalog No. 5000767480
Encompass
Aluminum foil masking discs are designed for applications requiring high-temperature resistance and precise masking. They provide a protective barrier and are ideal for powder coating, e-coating, and anodizing processes.
  • Made from durable aluminum foil
  • Silver finish for visual identification
  • Supplied on rolls for easy dispensing
  • Suitable for high-temperature masking applications
Catalog No. 50-007-67480 Supplier MSC Supplier No. 33510256
Aluminum foil masking discs are designed for applications requiring high-temperature resistance and precise masking. They provide a protective barrier and are ideal for powder coating, e-coating, and anodizing processes.
  • Made from durable aluminum foil
  • Silver finish for visual identification
  • Supplied on rolls for easy dispensing
  • Suitable for high-temperature masking applications