Learn More
Caplugs™ Evergreen™ CryoSure™ Vial Caps
Catalog No. 055586 Shop All Caplugs Products
Change view
Click to view available options
Color:
Green
Catalog No. Color
05-558-6 Green
1 options
Catalog No. 05-558-6 Supplier Caplugs™ Supplier No. 3003900G20
Add to Cart
Add to Cart
Screw caps to fit all sizes of Caplugs Evergreen™ CryoSure™ cyrogenic vials
  • Non-sterile
  • Green
  • 200/Pk.
Specifications
Color Green
Material Polyethylene
Sterility Non-sterile
Product Line CryoSure
Closure Type Screw Cap
Quantity 200
Autoclavable Not Autoclavable