Learn More
MSC Caplugs Finger Grip, 4-1/2 in, 7/8 in ID, 1 ea
Catalog No. 5000768071
Encompass
This protective grip is designed to provide a secure, finger-lugged handling solution for various industrial and laboratory applications. It features a durable construction, offering a firm, cushioned hold on cylindrical objects.
  • Finger lugged style for enhanced ergonomic grip
  • Black color for standard identification
  • Length of 4-1/2 in to accommodate diverse components
  • Internal diameter of 7/8 in for a snug fit
Catalog No. 50-007-68071 Supplier MSC Supplier No. 02847028
This protective grip is designed to provide a secure, finger-lugged handling solution for various industrial and laboratory applications. It features a durable construction, offering a firm, cushioned hold on cylindrical objects.
  • Finger lugged style for enhanced ergonomic grip
  • Black color for standard identification
  • Length of 4-1/2 in to accommodate diverse components
  • Internal diameter of 7/8 in for a snug fit