Learn More
MSC Caplugs High Temperature Masking Tape, 7.5 mil, 500 Pieces
Catalog No. 5000802417
Encompass
This Caplugs high temperature masking tape is designed for applications requiring heat resistance and clean removal. Made from 7.5 mil thick, off-white crepe paper with an acrylic adhesive, it is suitable for processes like stripping.
  • Maximum operating temperature up to 149°C (300°F)
  • Clean removal properties
  • Heat-resistant for demanding applications
  • Flake-resistant design
  • Durable construction
Catalog No. 50-008-02417 Supplier MSC Supplier No. 33509381
This Caplugs high temperature masking tape is designed for applications requiring heat resistance and clean removal. Made from 7.5 mil thick, off-white crepe paper with an acrylic adhesive, it is suitable for processes like stripping.
  • Maximum operating temperature up to 149°C (300°F)
  • Clean removal properties
  • Heat-resistant for demanding applications
  • Flake-resistant design
  • Durable construction